missing translation for 'onlineSavingsMsg'
Learn More

3-Carene, 90%, stabilized

Product Code. 10487212 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
25mL
This item is not returnable. View return policy

Product Code. 10487212

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13466-78-9 | C10H16 | 136.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13466-78-9, 138-86-3, 127-91-3, 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.23
MDL Number MFCD00001315
InChI Key BQOFWKZOCNGFEC-UHFFFAOYSA-N
Synonym 3-carene, carene, 3,7,7-trimethylbicyclo 4.1.0 hept-3-ene, delta-3-carene, monoterpenes, alpha-carene, bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl, +--delta3-carene, + car-3-ene, +--3-carene
PubChem CID 26049
ChEBI CHEBI:35661
IUPAC Name 4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CCC2C(C1)C2(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8640g/mL
Boiling Point 166°C to 170°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.474
Quantity 25 mL
Beilstein 05, 94
Merck Index 15, 1835
Specific Gravity 0.864
Solubility Information Solubility in water: insoluble
Stabilizer 0.01% alpha-Tocopherol
Formula Weight 136.23
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 3-Carene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.