missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-benzisothiazole, 97+%

Product Code. 11394755
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394755 5 g 5g
11304765 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394755 Supplier Thermo Scientific Alfa Aesar Supplier No. L17791.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7716-66-7 | C7H4ClNS | 169.626 g/mol

3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7716-66-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00673254
InChI Key BCPVKLRBQLRWDQ-UHFFFAOYSA-N
Synonym 3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
PubChem CID 598190
IUPAC Name 3-chloro-1,2-benzothiazole
SMILES C1=CC=C2C(=C1)C(=NS2)Cl

Specifications

Melting Point 37°C to 44°C
Boiling Point 80°C to 86°C (0.75 mmHg)
Quantity 5 g
Beilstein 119370
Solubility Information Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Formula Weight 169.63
Percent Purity 99%
Chemical Name or Material 3-Chloro-1,2-benzisothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.