missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1H-pyrazolo[3,4-b]pyridine, TRC

Product Code. 30365560
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365560 25 mg 25mg
30361881 100 mg 100mg
30360741 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365560 Supplier Toronto Research Chemicals Supplier No. TRCC38042525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117007-51-9
Molecular Formula C6 H4 Cl N3
Molecular Weight (g/mol) 153.57
IUPAC Name 3-chloro-1H-pyrazolo[3,4-b]pyridine
SMILES Clc1n[nH]c2ncccc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H4ClN3/c7-5-4-2-1-3-8-6(4)10-9-5/h1-3H,(H,8,9,10)
Formula Weight 153.0094
Chemical Name or Material 3-Chloro-1H-Pyrazolo[3,4-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.