missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4(3H,5H)-furandione, 97%

Product Code. 10346824 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10346824

Brand: Thermo Scientific Acros 399650100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4971-55-5 | C4H3ClO3 | 134.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4971-55-5
Molecular Formula C4H3ClO3
Molecular Weight (g/mol) 134.52
MDL Number MFCD00134273
InChI Key DJEJYZZJYYNADA-UHFFFAOYNA-N
Synonym 3-chloro-2,4 3h,5h-furandione, 3-chlorofuran-2,4 3h,5h-dione, 3-chloro-furan-2,4-dione, 3-chlorotetronic acid, 2,4 3h,5h-furandione, 3-chloro, 3-chloro-3,5-dihydrofuran-2,4-dione, acmc-209khp, 3-chlorofuran-2,4-dione, 3-chloranyloxolane-2,4-dione, 3-chlorotetrahydrofuran-2,4-dione
PubChem CID 4158575
IUPAC Name 3-chlorooxolane-2,4-dione
SMILES ClC1C(=O)COC1=O

Specifications

Melting Point 208°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 10 g
Formula Weight 134.52
Percent Purity 97%
Chemical Name or Material 3-Chloro-2, 4(3H, 5H)-furandione
compliance-icons
Product Identifier
  • 3-Chloro-2,4(3H,5H)-furandione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.