missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Product Code. 15492029
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492029 5 g 5g
15482029 1 g 1g
15469106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492029 Supplier Thermo Scientific Alfa Aesar Supplier No. H64723.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72537-17-8
Molecular Formula C6H2ClF4N
Molecular Weight (g/mol) 199.53
MDL Number MFCD00191918
InChI Key GDSROTVTTLUHCO-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine
PubChem CID 1268075
IUPAC Name 3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(Cl)C=C(C=N1)C(F)(F)F

Specifications

Melting Point 16°C to 20°C (decomposition)
Color Colorless
Density 1.524 g/mL
Boiling Point 50°C to 55°C (11 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.433
Quantity 5 g
UN Number UN1993
Formula Weight 199.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.