missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-fluorobenzyl Bromide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-2-fluorobenzyl Bromide |
| CAS | 85070-47-9 |
| Molecular Formula | C7H5BrClF |
| Quantity | 25 g |
| Synonym | 1-(Bromomethyl)-3-chloro-2-fluoro-benzene, 1-(Bromomethyl)-3-chloro-2-fluorobenzene |
| InChI Formula | InChI=1S/C7H5BrClF/c8-4-5-2-1-3-6(9)7(5)10/h1-3H,4H2 |
| SMILES | Fc1c(Cl)cccc1CBr |
| IUPAC Name | 1-(bromomethyl)-3-chloro-2-fluorobenzene |
| Molecular Weight (g/mol) | 223.47 |
| Formula Weight | 221.9247 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction