missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoropyridine, 95%, Thermo Scientific™

Product Code. 15493767
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493767 250 mg 250mg
11312919 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27755.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1480-64-4
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04114144
InChI Key IHGMHTQDGNVKTA-UHFFFAOYSA-N
PubChem CID 2762818
IUPAC Name 3-chloro-2-fluoropyridine
SMILES C1=CC(=C(N=C1)F)Cl

Specifications

Density 1.33
Boiling Point 162°C
Flash Point 52°C (126°F)
Odor Unpleasant
Refractive Index 1.503
Quantity 250 mg
UN Number UN2924
Synonym 3-chloro-2-fluoro-pyridine, 2-fluoro-3-chloropyridine, pyridine, 3-chloro-2-fluoro, chlorofluoropyridine, pubchem6263, acmc-1byou, 3-chloro-2-fluoropyridine, ksc525m9d, pyridine,3-chloro-2-fluoro, 3-chloro-2-fluoro pyridine
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-fluoropyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.