missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylpropene, 90%, tech.

Product Code. 10031580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031580 50 mL Glass bottle 50mL
10457920 250 mL Glass bottle 250mL
10447920 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031580 Supplier Thermo Scientific Acros Supplier No. 148420500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 563-47-3 | C4H7Cl | 90.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-47-3
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000953
InChI Key OHXAOPZTJOUYKM-UHFFFAOYSA-N
Synonym 3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile
PubChem CID 11241
ChEBI CHEBI:82419
IUPAC Name 3-chloro-2-methylprop-1-ene
SMILES CC(=C)CCl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point -80°C
Density 0.9200g/mL
Boiling Point 71°C to 72°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CH2Cl
Refractive Index 1.4262 to 1.4282
Quantity 50 mL
Beilstein 01, 209
Merck Index 15, 2150
Specific Gravity 0.92
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 0.42 mPa.s (20°C)
Formula Weight 90.55
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methylpropene, tech., 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-2-methylpropene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.