missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-3-methyl-1-butyne, 95%

Product Code. 11470788
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11470788 1 g 1g
11480788 5 g 5g
11490788 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11470788

Brand: Thermo Scientific Alfa Aesar B25252.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-97-3 | C5H7Cl | 102.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-97-3
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.56
MDL Number MFCD00190221
InChI Key QSILYWCNPOLKPN-UHFFFAOYSA-N
Synonym 3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
PubChem CID 70679
IUPAC Name 3-chloro-3-methylbut-1-yne
SMILES CC(C)(Cl)C#C

Specifications

Melting Point -61°C
Boiling Point 73°C to 75°C
Flash Point 4°C (40°F)
Odor Recognizable
Quantity 1 g
UN Number UN1993
Formula Weight 102.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-3-methyl-1-butyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.