missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-3-methyl-1-butyne, 96%

Product Code. 10558463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10558463 25 g Glass bottle 25g
10220023 1 g Glass bottle 1g
10630545 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10558463

Brand: Thermo Scientific Acros 378010250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-97-3 | C5H7Cl | 102.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-97-3
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.56
MDL Number MFCD00190221
InChI Key QSILYWCNPOLKPN-UHFFFAOYSA-N
Synonym 3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
PubChem CID 70679
IUPAC Name 3-chloro-3-methylbut-1-yne
SMILES CC(C)(Cl)C#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Straw
Density 0.9100g/mL
Boiling Point 73°C to 75°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.419
Quantity 25 g
Specific Gravity 0.91
Formula Weight 102.56
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloro-3-methyl-1-butyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-3-methyl-1-butyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.