missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10642305
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10642305 25 g 25g
10671313 1 g 1g
10475142 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10642305 Supplier Thermo Scientific Maybridge Supplier No. TL00685EB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 42413-03-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD00203987
InChI Key GNYVVCRRZRVBDD-UHFFFAOYSA-N
Synonym 3-chloro-4-methylbenzene-1-sulfonyl chloride, 3-chloro-4-methylbenzenesulphonyl chloride, pubchem5098, acmc-209jpd, 2-chloro-p-toluenesulfonyl chloride, chloro 3-chloro-4-methylphenyl sulfone, 3-chloro-4-methylbenzenesulphonylchloride, 3-chloro-4-methylbenzensulfonyl chloride, 3-chloro-4-methylphenylsulfonyl chloride, 2-chloro-toluene-4-sulfonic acid chloride
PubChem CID 259744
IUPAC Name 3-chloro-4-methylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methylbenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.