missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-4'-(trifluoromethoxy)acetophenone, 98%, Thermo Scientific™

Product Code. 15468338
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468338 5 g 5g
15426766 250 mg 250mg
15458338 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468338 Supplier Thermo Scientific Alfa Aesar Supplier No. H61768.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3′-Chloro-4′-(trifluoromethoxy)acetophenone is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129604-27-9
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD12545820
InChI Key BUSQQCHYIYPNTO-UHFFFAOYSA-N
Synonym 1-3-chloro-4-trifluoromethoxy phenyl ethanone, 3-chloro-4-trifluoromethoxy acetophenone, 3'-chloro-4'-trifluoromethoxyacetophenone, 3'-chloro-4'-trifluoromethoxy acetophenone, intermediates-zcf02001, ethanone, 1-3-chloro-4-trifluoromethoxy phenyl, 1-3-chloro-4-trifluoromethoxy phenyl ethan-1-one
PubChem CID 14793273
IUPAC Name 1-[3-chloro-4-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC(F)(F)F)Cl

Specifications

Color Colorless
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 238.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3'-Chloro-4'-(trifluoromethoxy)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.