missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(3-chloropropyl)-10,11-dihydro-5H-dibenz[b,f]azepine (>90%), TRC

Product Code. 30364481
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364481 100 mg 100mg
30351076 1 g 1g
30351077 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364481 Supplier Toronto Research Chemicals Supplier No. TRCC365075100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51551-41-8
Molecular Formula C17 H17 Cl2 N
Molecular Weight (g/mol) 306.23
IUPAC Name 2-chloro-11-(3-chloropropyl)-5,6-dihydrobenzo[b][1]benzazepine
SMILES ClCCCN1c2ccccc2CCc3ccc(Cl)cc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H17Cl2N/c18-10-3-11-20-16-5-2-1-4-13(16)6-7-14-8-9-15(19)12-17(14)20/h1-2,4-5,8-9,12H,3,6-7,10-11H2
Formula Weight 305.0738
Chemical Name or Material 3-Chloro-5-(3-chloropropyl)-10,11-dihydro-5H-dibenz[b,f]azep
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.