missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, TRC

Product Code. 30351512
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351512 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351512 Supplier Toronto Research Chemicals Supplier No. TRCC432835100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80194-68-9
Molecular Formula C7 H3 Cl F3 N O2
Quantity 100 mg
Synonym 3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid, 3-Chloro-5-(trifluoromethyl)pyridin-2-carboxylic acid, 3-Chloro-5-trifluoromethyl-pyridine-2-carboxylic acid
InChI Formula InChI=1S/C7H3ClF3NO2/c8-4-1-3(7(9,10)11)2-12-5(4)6(13)14/h1-2H,(H,13,14)
SMILES OC(=O)c1ncc(cc1Cl)C(F)(F)F
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol) 225.55
Formula Weight 224.9804
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)picolinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.