missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 15435097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15435097 5 g 5g
15425097 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15435097

Brand: Thermo Scientific Alfa Aesar H32376.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85148-26-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042227
InChI Key OMRCXTBFBBWTDL-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine, 3-trifluoromethyl-5-chloropyridine, pyridine, 3-chloro-5-trifluoromethyl, pubchem3552, acmc-209zee, ksc495c1f, 3-chloro-5-trifluoromethyl-pyridine, 3-chloro-5-trifluoromethyl pyridine-carbonitrile
PubChem CID 2736698
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=CN=C1

Specifications

Boiling Point 50°C to 51°C (4 mmHg)
Quantity 5 g
UN Number UN2810
Formula Weight 181.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.