missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-hydrazinopyridazine, 98%

Product Code. 11392386
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392386 1 g 1g
11302396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392386 Supplier Thermo Scientific Alfa Aesar Supplier No. L06121.03
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17284-97-8 | C4H5ClN4 | 144.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17284-97-8
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.562
MDL Number MFCD00051740
InChI Key FXYQRYGWWZKUFV-UHFFFAOYSA-N
Synonym 3-chloro-6-hydrazinopyridazine, 3-chloro-6-hydrazinylpyridazine, 3-chloropyridazin-6-yl hydrazine, 1-6-chloropyridazin-3-yl hydrazine, 6-chloro-pyridazin-3-yl-hydrazine, 3-chloro-6-hydrazino-pyridazine, 6-chloropyridazin-3-yl hydrazine, 3 2h-pyridazinone, 6-chloro-, hydrazone, 6-chloropyridazine-3-ylhydrazine, 6-chloro-3-pyridazinyl hydrazine
PubChem CID 100787
IUPAC Name (6-chloropyridazin-3-yl)hydrazine
SMILES C1=CC(=NN=C1NN)Cl

Specifications

Melting Point 134°C to 140°C
Quantity 1 g
Beilstein 125591
Formula Weight 144.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-6-hydrazinopyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.