missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-iodopyridazine, 95%

Product Code. 15400829
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400829 1 g 1g
15410829 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400829 Supplier Thermo Scientific Alfa Aesar Supplier No. H63879.03
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 135034-10-5 | C4H2ClIN2 | 240.43 g/mol

Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135034-10-5
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.43
MDL Number MFCD08275187
InChI Key PNEPCDPKMXJYIQ-UHFFFAOYSA-N
Synonym 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine
PubChem CID 15418839
IUPAC Name 3-chloro-6-iodopyridazine
SMILES ClC1=NN=C(I)C=C1

Specifications

Melting Point 110°C to 120°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 240.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Chloro-6-iodopyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.