missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-phenylpyridazine, TRC

Product Code. 30003301
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003301 50 mg 50mg
30008166 10 mg 10mg
30007777 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003301 Supplier Toronto Research Chemicals Supplier No. TRCC59667350MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20375-65-9
Molecular Formula C10H7N2Cl
Molecular Weight (g/mol) 190.63
Synonym 6-Phenyl-3-chloropyridazine, NSC 28739, 3-Phenyl-6-chloropyridazine, 6-Chloro-3-phenylpyridazine
IUPAC Name 3-chloro-6-phenylpyridazine
SMILES Clc1ccc(nn1)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H7ClN2/c11-10-7-6-9(12-13-10)8-4-2-1-3-5-8/h1-7H
Formula Weight 190.0298
Chemical Name or Material 3-Chloro-6-phenylpyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.