missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-alpha-toluenesulfonyl chloride, 96%

Product Code. 11421067
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11421067

Brand: Thermo Scientific Alfa Aesar B24073.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24974-73-0 | C7H6Cl2O2S | 225.083 g/mol

3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24974-73-0
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD02683111
InChI Key LXTGNVLBPVVMSL-UHFFFAOYSA-N
Synonym 3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro
PubChem CID 2757802
IUPAC Name (3-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl

Specifications

Melting Point 69°C to 75°C
Quantity 1 g
UN Number UN3261
Formula Weight 225.09
Percent Purity 96%
Chemical Name or Material 3-Chloro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.