missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-alpha-toluenesulfonyl chloride, 96%

Product Code. 11441067
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441067 25 g 25g
11421067 1 g 1g
11431067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441067 Supplier Thermo Scientific Alfa Aesar Supplier No. B24073.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24974-73-0 | C7H6Cl2O2S | 225.083 g/mol

3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24974-73-0
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD02683111
InChI Key LXTGNVLBPVVMSL-UHFFFAOYSA-N
Synonym 3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro
PubChem CID 2757802
IUPAC Name (3-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl

Specifications

Melting Point 69°C to 75°C
Quantity 25 g
UN Number UN3261
Formula Weight 225.09
Percent Purity 96%
Chemical Name or Material 3-Chloro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.