missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzamide, 98%, ACROS Organics™

Product Code. 10031751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031751 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10031751 Supplier Thermo Scientific Acros Supplier No. 351370250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 133°C to 137°C
CAS 618-48-4
CAS Min % 97.5
Color White
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 98%
Molecular Formula C7H6ClNO
Linear Formula ClC6H4C(O)NH2
MDL Number MFCD00014799
Synonym m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce
InChI Key MJTGQALMWUUPQM-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=CC(Cl)=C1
IUPAC Name 3-chlorobenzamide
Molecular Weight (g/mol) 155.58
PubChem CID 69254
ChEBI CHEBI:10587
Formula Weight 155.58
Percent Purity ≥97.5%
Chemical Name or Material 3-Chlorobenzamide, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.