missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10620476
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10620476 25 g 25g
10602243 1 g 1g
10552432 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10620476 Supplier Thermo Scientific Maybridge Supplier No. BTB00300EB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21815-91-8
Molecular Formula C9H4Cl2OS
Molecular Weight (g/mol) 231.09
MDL Number MFCD00053069
InChI Key GWKSSMDJEWPKCM-UHFFFAOYSA-N
Synonym 3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride
PubChem CID 519898
IUPAC Name 3-chloro-1-benzothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=C(Cl)C2=CC=CC=C2S1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 231.1
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.