missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzonitrile, 99%

Product Code. 11480763
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480763 50 g 50g
11470763 10 g 10g
11490763 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480763 Supplier Thermo Scientific Alfa Aesar Supplier No. A13237.18
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 766-84-7 | C7H4ClN | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-84-7
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001798
InChI Key WBUOVKBZJOIOAE-UHFFFAOYSA-N
Synonym m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
PubChem CID 13015
IUPAC Name 3-chlorobenzonitrile
SMILES ClC1=CC=CC(=C1)C#N

Specifications

Melting Point 39°C to 42°C
Density 1.14
Quantity 50 g
Odor Almond-like
UN Number UN3439
Beilstein 1906694
Boiling Point 94°C (10 mmHg)
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 3-Chlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.