missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10366854
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10366854 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366854 Supplier Thermo Scientific Maybridge Supplier No. CC46524CB
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 869901-13-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H11ClN2
MDL Number MFCD08271938
Quantity 250 mg
Synonym 3-chloromethyl-1-methyl-5-phenyl-1h-pyrazole, 3-chloromethyl-1-methyl-5-phenylpyrazole, 1h-pyrazole,3-chloromethyl-1-methyl-5-phenyl
InChI Key KAZKAWDJSYXNGK-UHFFFAOYSA-N
SMILES CN1C(=CC(=N1)CCl)C2=CC=CC=C2
IUPAC Name 3-(chloromethyl)-1-methyl-5-phenylpyrazole
Molecular Weight (g/mol) 206.673
PubChem CID 18525795
Formula Weight 206.67
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.