missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole, 95%, Thermo Scientific™

Product Code. 10265324
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10265324 1 g 1g
11330647 250 mg 250mg
11340647 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265324 Supplier Thermo Scientific Maybridge Supplier No. CC46924DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-48-2
Molecular Formula C9H9ClN2O
Molecular Weight (g/mol) 196.63
MDL Number MFCD08690269
InChI Key DZUIHVJDKDNADW-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-2-furyl-1-methyl-1h-pyrazole, 3-chloromethyl-5-furan-2-yl-1-methylpyrazole, 1h-pyrazole,3-chloromethyl-5-2-furanyl-1-methyl, 3-chloromethyl-5-furan-2-yl-1-methyl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl furan
PubChem CID 18525803
IUPAC Name 3-(chloromethyl)-5-(furan-2-yl)-1-methylpyrazole
SMILES CN1N=C(CCl)C=C1C1=CC=CO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 196.64
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.