missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-furyl)isoxazole, ≥97%, Thermo Scientific™

Product Code. 10259772
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10259772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10259772 Supplier Thermo Scientific Maybridge Supplier No. CC34924DA
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 848658-70-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H6ClNO2
MDL Number MFCD07329988
Quantity 1 g
Synonym 3-chloromethyl-5-2-furyl isoxazole, 3-chloromethyl-5-furan-2-yl-1,2-oxazole, isoxazole,3-chloromethyl-5-2-furanyl, 3-chloromethyl-5-furan-2-yl isoxazole
InChI Key IPIMDVOPXIOKAK-UHFFFAOYSA-N
SMILES C1=COC(=C1)C2=CC(=NO2)CCl
IUPAC Name 3-(chloromethyl)-5-(furan-2-yl)-1,2-oxazole
Molecular Weight (g/mol) 183.591
PubChem CID 4962799
Formula Weight 183.59
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-(2-furyl)isoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.