missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10613043
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10613043 250 mg 250mg
10497632 1 g 1g
10082214 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10613043 Supplier Thermo Scientific Maybridge Supplier No. SEW06262CB
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-06-1
Molecular Formula C7H5ClN2OS
Molecular Weight (g/mol) 200.64
MDL Number MFCD01571188
InChI Key YVYZVJRSESVBCM-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-thiophen-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-2-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thiophen-2-yl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-2-thienyl, 2-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
PubChem CID 2798343
IUPAC Name 3-(chloromethyl)-5-thiophen-2-yl-1,2,4-oxadiazole
SMILES C1=CSC(=C1)C2=NC(=NO2)CCl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.65
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.