missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloroperbenzoic Acid (Technical Grade, wet with water), TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3-Chloroperbenzoic Acid (Technical Grade) |
| CAS | 937-14-4 |
| Molecular Formula | C7H5ClO3 |
| Quantity | 25 g |
| Synonym | 3-Chlorobenzenecarboperoxoic Acid, m-Chloroperoxybenzoic Acid, 3-Chlorobenzenecarboperoxoic Acid, 3-Chloroperbenzoic Acid, 3-Chloroperoxybenzoic Acid, MCPBA, NSC 97094, m-Chlorobenzoyl Hydroperoxide, m-Chloroperbenzoic Acid, m-Chloroperoxobenzoic Acid, m-Chloroperoxybenzoic Acid, m-Chloroperoxybenzoic Acid |
| InChI Formula | InChI=1S/C7H5ClO3/c8-6-3-1-2-5(4-6)7(9)11-10/h1-4,10H |
| SMILES | OOC(=O)c1cccc(Cl)c1 |
| IUPAC Name | 3-chlorobenzenecarboperoxoic acid |
| Molecular Weight (g/mol) | 172.57 |
| Formula Weight | 171.9927 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction