missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroperbenzoic Acid (Technical Grade, wet with water), TRC

Product Code. 30362481
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30362481

Brand: Toronto Research Chemicals TRCC37491550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloroperbenzoic Acid (Technical Grade)
CAS 937-14-4
Molecular Formula C7H5ClO3
Quantity 50 g
Synonym 3-Chlorobenzenecarboperoxoic Acid, m-Chloroperoxybenzoic Acid, 3-Chlorobenzenecarboperoxoic Acid, 3-Chloroperbenzoic Acid, 3-Chloroperoxybenzoic Acid, MCPBA, NSC 97094, m-Chlorobenzoyl Hydroperoxide, m-Chloroperbenzoic Acid, m-Chloroperoxobenzoic Acid, m-Chloroperoxybenzoic Acid, m-Chloroperoxybenzoic Acid
InChI Formula InChI=1S/C7H5ClO3/c8-6-3-1-2-5(4-6)7(9)11-10/h1-4,10H
SMILES OOC(=O)c1cccc(Cl)c1
IUPAC Name 3-chlorobenzenecarboperoxoic acid
Molecular Weight (g/mol) 172.57
Formula Weight 171.9927
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.