missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenethylalcohol, 98%, Thermo Scientific™

Product Code. 10632983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10632983 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10632983

Brand: Thermo Scientific Acros 185960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5182-44-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00002892
InChI Key NDWAVJKRSASRPH-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
PubChem CID 78856
SMILES OCCC1=CC=CC(Cl)=C1

Specifications

CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.1810g/mL
Boiling Point 135.0°C to 137.0°C (13.0 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5475 to 1.5495
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.181
Linear Formula ClC6H4CH2CH2OH
IUPAC Name 2-(3-chlorophenyl)ethanol
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 3-Chlorophenethylalcohol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.