missing translation for 'onlineSavingsMsg'
Learn More

(3-Chlorophenoxy)acetyl chloride, 95%

Product Code. 11334995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334995 5 g 5g
11324995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334995 Supplier Thermo Scientific Alfa Aesar Supplier No. L18427.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 114476-84-5 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114476-84-5
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD02094018
InChI Key KPINZNVDHXIYAR-UHFFFAOYSA-N
Synonym 3-chlorophenoxyacetyl chloride, 3-chlorophenoxy acetyl chloride, 2-3-chlorophenoxy acetyl chloride, acetyl chloride, 3-chlorophenoxy, acmc-1brdk, acetyl chloride,2-3-chlorophenoxy
PubChem CID 7010339
IUPAC Name 2-(3-chlorophenoxy)acetyl chloride
SMILES C1=CC(=CC(=C1)Cl)OCC(=O)Cl

Specifications

Boiling Point 120°C to 122°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 5 g
UN Number UN3265
Beilstein 1871611
Sensitivity Moisture sensitive
Formula Weight 205.04
Percent Purity 95%
Chemical Name or Material (3-Chlorophenoxy)acetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.