missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Product Code. 10526114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10526114 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10526114 Supplier Thermo Scientific Acros Supplier No. 397491000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36229-42-2 | C6H4BrClMg | 215.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36229-42-2, 109-99-9
CAS Min % 11.24, 86.52
CAS Max % 13.48, 88.76
Quantity 100 mL
Molecular Formula C6H4BrClMg
Linear Formula ClC6H4MgBr
MDL Number MFCD00672003
Flash Point <−17°C
Synonym 3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key ZIYRUWPTUPFRQU-UHFFFAOYSA-M
SMILES ClC1=CC([Mg]Br)=CC=C1
IUPAC Name magnesium;chlorobenzene;bromide
Molecular Weight (g/mol) 215.76
PubChem CID 12252967
Formula Weight 215.76
Concentration or Composition (by Analyte or Components) 0.5M (Argentometry), exact strength on the certificate of analysis
Identification Identity confirmed
Packaging AcroSeal™ Glass Bottle
Appearance Clear yellow to brown solution
Specific Gravity 0.96
Density 0.9600g/mL
Chemical Name or Material 3-Chlorophenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chlorophenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.