missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15445405
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445405 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445405 Supplier Thermo Scientific Alfa Aesar Supplier No. H58709.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Spezifikation

CAS 186000-42-0
Density 1.032
Molecular Formula C6H5ClIZn-
MDL Number MFCD01311393
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 3-chlorophenylzinc iodide
InChI Key RZAFMWURTCHXIE-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)Cl.[Zn].[I-]
IUPAC Name chlorobenzene;zinc;iodide
Molecular Weight (g/mol) 304.84
PubChem CID 131668582
Formula Weight 303.84
Chemical Name or Material 3-Chlorophenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.