missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropanesulfonyl chloride, 96%

Código de producto. 10675202 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Tamaño de la unidad:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging Tamaño de la unidad
10675202 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10675202 Proveedor Thermo Scientific Acros N.º de proveedor 154730250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1633-82-5 | C3H6Cl2O2S | 177.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 1633-82-5
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.4500g/mL
Boiling Point 70°C (0.5 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C3H6Cl2O2S
Linear Formula Cl(CH2)3SO2Cl
Refractive Index 1.4890 to 1.491
MDL Number MFCD00007463
Quantity 25 mL
Specific Gravity 1.45
Synonym 3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
Solubility Information Solubility in water: reacts
InChI Key GPKDGVXBXQTHRY-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)Cl)CCl
IUPAC Name 3-chloropropane-1-sulfonyl chloride
Molecular Weight (g/mol) 177.05
PubChem CID 15410
Formula Weight 177.05
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloropropanesulfonyl chloride, 96%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 3-Chloropropanesulfonyl chloride
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.