missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropanesulfonyl chloride, 98+%

Product Code. 11423354
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423354 50 g 50g
11413354 10 g 10g
11433354 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423354 Supplier Thermo Scientific Alfa Aesar Supplier No. A10631.18
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1633-82-5 | C3H6Cl2O2S | 177.039 g/mol

3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.

Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-82-5
Molecular Formula C3H6Cl2O2S
Molecular Weight (g/mol) 177.039
MDL Number MFCD00007463
InChI Key GPKDGVXBXQTHRY-UHFFFAOYSA-N
Synonym 3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
PubChem CID 15410
IUPAC Name 3-chloropropane-1-sulfonyl chloride
SMILES C(CS(=O)(=O)Cl)CCl

Specifications

Density 1.456
Boiling Point 70°C (0.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula Cl(CH2)3SO2Cl
Refractive Index 1.49
Quantity 50 g
UN Number UN3265
Beilstein 1754119
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ethyl Acetate and hexane.
Formula Weight 177.05
Percent Purity ≥98%
Chemical Name or Material 3-Chloropropanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.