missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropanesulfonyl chloride, 98+%

Product Code. 11433354
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Tamaño de la unidad:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
11433354 250 g 250g
11413354 10 g 10g
11423354 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11433354 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A10631.30
 Restricted Item
Estimated Shipment: 25-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1633-82-5 | C3H6Cl2O2S | 177.039 g/mol

3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.

Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1633-82-5
Molecular Formula C3H6Cl2O2S
Molecular Weight (g/mol) 177.039
MDL Number MFCD00007463
InChI Key GPKDGVXBXQTHRY-UHFFFAOYSA-N
Synonym 3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
PubChem CID 15410
IUPAC Name 3-chloropropane-1-sulfonyl chloride
SMILES C(CS(=O)(=O)Cl)CCl

Especificaciones

Density 1.456
Boiling Point 70°C (0.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula Cl(CH2)3SO2Cl
Refractive Index 1.49
Quantity 250 g
UN Number UN3265
Beilstein 1754119
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ethyl Acetate and hexane.
Formula Weight 177.05
Percent Purity ≥98%
Chemical Name or Material 3-Chloropropanesulfonyl chloride
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.