missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionitrile, 98%

Product Code. 11377625
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377625 25 g 25g
11387625 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377625 Supplier Thermo Scientific Alfa Aesar Supplier No. L05902.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 542-76-7 | C3H4ClN | 89.52 g/mol

3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-76-7
Molecular Formula C3H4ClN
Molecular Weight (g/mol) 89.52
MDL Number MFCD00001952
InChI Key GNHMRTZZNHZDDM-UHFFFAOYSA-N
Synonym 3-chloropropionitrile, propanenitrile, 3-chloro, 3-chloropropiononitrile, 1-chloro-2-cyanoethane, 3-chloropropanonitrile, 3-chlorpropannitril, propionitrile, 3-chloro, beta-chloropropionitrile, usaf a-8798, rcra waste number p027
PubChem CID 10963
IUPAC Name 3-chloropropanenitrile
SMILES ClCCC#N

Specifications

Melting Point -51°C
Density 1.143
Boiling Point 174°C to 176°C
Flash Point 75°C (167°F)
Refractive Index 1.438
Quantity 25 g
UN Number UN3276
Beilstein 1098454
Merck Index 14,2160
Solubility Information Soluble in water. (4.5g/100mL,25°C). Miscible in Ether,Alcohols,Benzene,Acetone.
Formula Weight 89.53
Percent Purity 98%
Chemical Name or Material 3-Chloropropionitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.