missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11461727
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461727 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461727 Supplier Thermo Scientific Alfa Aesar Supplier No. B25698.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ mois
TRUSTED_SUSTAINABILITY

Specifications

CAS 625-36-5
Melting Point -32°C
Density 1.325
Boiling Point 143°C to 145°C
Flash Point 59°C
Molecular Formula C3H4Cl2O
Linear Formula ClCH2CH2COCl
Refractive Index 1.457
MDL Number MFCD00000747
Quantity 500 g
UN Number UN3389
Beilstein 635814
Sensitivity Moisture sensitive
Synonym 3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
Solubility Information Reacts violently with water.
InChI Key INUNLMUAPJVRME-UHFFFAOYSA-N
SMILES ClCCC(Cl)=O
IUPAC Name 3-chloropropanoyl chloride
Molecular Weight (g/mol) 126.96
PubChem CID 69364
Formula Weight 126.97
Percent Purity 97%
Chemical Name or Material 3-Chloropropionyl chloride
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.