missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)benzene, 98%

Product Code. 11454407
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454407 5 g 5g
11464407 25 g 25g
11474407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454407 Supplier Thermo Scientific Alfa Aesar Supplier No. B20445.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104-52-9 | C9H11Cl | 154.637 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-52-9
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00001001
InChI Key XZBXAYCCBFTQHH-UHFFFAOYSA-N
Synonym 3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
PubChem CID 7706
IUPAC Name 3-chloropropylbenzene
SMILES C1=CC=C(C=C1)CCCCl

Specifications

Density 1.05
Boiling Point 218°C to 220°C
Flash Point 87°C (189°F)
Linear Formula C6H5(CH2)3Cl
Refractive Index 1.522
Quantity 5 g
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material (3-Chloropropyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.