missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl isocyanate, 96%, Thermo Scientific™

Product Code. p-91535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10668634 1 g Glass bottle 1g
10368790 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10668634 Supplier Thermo Scientific Acros Supplier No. 314590010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13010-19-0
Molecular Formula C4H6ClNO
Molecular Weight (g/mol) 119.55
MDL Number MFCD00010384
InChI Key RQAVSDINDRNIKL-UHFFFAOYSA-N
Synonym 3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
PubChem CID 166752
IUPAC Name 1-chloro-3-isocyanatopropane
SMILES ClCCCN=C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.1600g/mL
Boiling Point 152°C to 154°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3NCO
Refractive Index 1.4490 to 1.453
Quantity 1 g
Specific Gravity 1.16
Formula Weight 119.55
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloropropyl isocyanate, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.