missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trimethylsilane, 98%

Product Code. 11371587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371587 5 g 5g
11381587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371587 Supplier Thermo Scientific Alfa Aesar Supplier No. L04458.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2344-83-4 | C6H15ClSi | 150.72 g/mol

(3-Chloropropyl)trimethylsilane used in the preparation of: 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions, 5-trimethylsilyl pentanol, N-(tri-methylsilylalkyl)diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Chloropropyl)trimethylsilane used in the preparation of: 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions, 5-trimethylsilyl pentanol, N-(tri-methylsilylalkyl)diamines.

Solubility
Not miscible in water.

Notes
Store under inert gas. Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2344-83-4
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00045299
InChI Key QXDDDCNYAAJLBT-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethylsilane, silane, 3-chloropropyl trimethyl, 3-chloropropyl trimethyl silane, acmc-20apo8, chloropropyltrimethylsilane, chloropropyl trimethyl silane, 3 3-chloropropyl trimethylsilane, gamma-chloropropyl trimethylsilane
PubChem CID 75362
IUPAC Name 3-chloropropyl(trimethyl)silane
SMILES C[Si](C)(C)CCCCl

Specifications

Boiling Point 150°C to 152°C
Quantity 5 g
UN Number UN1993
Beilstein 1732625
Flash Point 49°C (120°F)
Solubility Information Not miscible in water.
Formula Weight 150.73
Percent Purity 98%
Refractive Index 1.432
Density 0.879
Chemical Name or Material (3-Chloropropyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.