missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropylamine hydrochloride, 98%

Product Code. 11468396
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468396 250 g 250g
11458396 50 g 50g
11448396 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468396 Supplier Thermo Scientific Alfa Aesar Supplier No. A15415.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6276-54-6 | C3H9Cl2N | 130.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6276-54-6
Molecular Formula C3H9Cl2N
Molecular Weight (g/mol) 130.012
MDL Number MFCD00012913
InChI Key IHPRVZKJZGXTBQ-UHFFFAOYSA-N
Synonym 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
PubChem CID 11469095
IUPAC Name 3-chloropropan-1-amine;hydrochloride
SMILES C(CN)CCl.Cl

Specifications

Melting Point 147°C to 152°C
Quantity 250 g
Beilstein 3667451
Formula Weight 130.02
Percent Purity 98%
Chemical Name or Material 3-Chloropropylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.