missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyridine, 99%

Product Code. 11415747
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11415747

Brand: Thermo Scientific Alfa Aesar B24316.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-60-8 | C5H4ClN | 113.54 g/mol

3-Chloropyridine is used as a pharmaceutical intermediate. It can be used to produce 3-chloro-pyridine-1-oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropyridine is used as a pharmaceutical intermediate. It can be used to produce 3-chloro-pyridine-1-oxide.

Solubility
Soluble in water 10 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-60-8
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.54
MDL Number MFCD00006375
InChI Key PWRBCZZQRRPXAB-UHFFFAOYSA-N
Synonym pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine
PubChem CID 12287
IUPAC Name 3-chloropyridine
SMILES ClC1=CC=CN=C1

Specifications

Density 1.205
Boiling Point 147°C to 149°C
Flash Point 53°C (127°F)
Refractive Index 1.533
Quantity 50 g
UN Number UN1993
Beilstein 105744
Solubility Information Soluble in water 10g/L.
Formula Weight 113.55
Percent Purity 99%
Chemical Name or Material 3-Chloropyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.