missing translation for 'onlineSavingsMsg'
Learn More

3-(Chlorosulfonyl)benzoic acid, 96%

Product Code. 11416156
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416156 25 g 25g
11406156 5 g 5g
11426156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416156 Supplier Thermo Scientific Alfa Aesar Supplier No. B22987.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4025-64-3 | C7H5ClO4S | 220.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4025-64-3
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.62
MDL Number MFCD00024877
InChI Key LMRKXSDOAFUINK-UHFFFAOYSA-N
Synonym 3-chlorosulfonyl benzoic acid, benzoic acid, 3-chlorosulfonyl, 3-carboxybenzenesulfonyl chloride, m-chlorosulfonylbenzoic acid, 3-carboxyphenylsulfonyl chloride, benzoic acid, m-chlorosulfonyl, 3-chlorosulphonyl benzoic acid, 3-carboxylbenzenesulfonyl chloride, 3-chlorosulfonyl-benzoic acid, m-chlorosulfonyl benzoic acid
PubChem CID 77641
SMILES OC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 133°C to 138°C
Quantity 25 g
UN Number UN3261
Beilstein 2645465
Sensitivity Moisture sensitive
IUPAC Name 3-chlorosulfonylbenzoic acid
Formula Weight 220.63
Percent Purity 97%
Chemical Name or Material 3-(Chlorosulfonyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.