missing translation for 'onlineSavingsMsg'
Learn More

3-(Chlorosulfonyl)benzoyl chloride, 98%

Product Code. 10558702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10558702 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10558702 Supplier Thermo Scientific Acros Supplier No. 405070250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4052-92-0 | C7H4Cl2O3S | 239.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4052-92-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C to 20°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4Cl2O3S
Linear Formula COClC6H4SO2Cl
Refractive Index 1.5780 to 1.58
MDL Number MFCD00051352
Quantity 25 g
Synonym 3-chlorosulfonyl benzoyl chloride, benzoyl chloride, 3-chlorosulfonyl, m-chlorosulfonyl benzoyl chloride, 3-chlorosulphonyl benzoyl chloride, 3-chlorosulphonylbenzoyl chloride, 3-chlorosulfonyl-benzoyl chloride, benzoyl chloride,3-chlorosulfonyl, benzoyl chloride, m-chlorosulfonyl-7ci,8ci, c7h4cl2so3, 3-chlorosulfonylbenzoyl=chloride
InChI Key XWEBTVZIZWEJOO-UHFFFAOYSA-N
SMILES ClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
IUPAC Name 3-chlorosulfonylbenzoyl chloride
Molecular Weight (g/mol) 239.07
PubChem CID 77671
Formula Weight 239.08
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 3-(Chlorosulfonyl)benzoyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(Chlorosulfonyl)benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.