missing translation for 'onlineSavingsMsg'
Learn More

3-Chromanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10326864
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10326864 10 g Amber Glass Bottle 10g
10212982 250 mg Amber Glass Bottle 250mg
10273022 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10326864 Supplier Thermo Scientific Maybridge Supplier No. CC25801EA
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 115822-57-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD03783554
InChI Key UGAGZMGJJFSKQM-UHFFFAOYSA-N
Synonym chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid
PubChem CID 2795458
IUPAC Name 3,4-dihydro-2H-chromene-3-carboxylic acid
SMILES C1C(COC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material 3-Chromanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.