missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-1-propylboronic acid pinacol ester, 96%

Product Code. 11426230
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426230 5 g 5g
11416230 1 g 1g
11436230 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426230 Supplier Thermo Scientific Alfa Aesar Supplier No. H27126.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 238088-16-9 | C10H18BNO2 | 195.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 238088-16-9
Molecular Formula C10H18BNO2
Molecular Weight (g/mol) 195.07
MDL Number MFCD09953480
InChI Key LVMSRWXPZLRTIC-UHFFFAOYSA-N
Synonym 3-cyano-1-propylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 4-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 1,3,2-dioxaborolane-2-butanenitrile, 4,4,5,5-tetramethyl, 3-cyanoprop-1-ylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butyronitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl butanenitrile
PubChem CID 11183176
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile
SMILES CC1(C)OB(CCCC#N)OC1(C)C

Specifications

Refractive Index 1.437
Quantity 5 g
UN Number UN3276
Formula Weight 195.07
Percent Purity 96%
Chemical Name or Material 3-Cyano-1-propylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.