missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%

Product Code. 11454064
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454064 100 g 100g
11439713 5 g 5g
11449713 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454064 Supplier Thermo Scientific Alfa Aesar Supplier No. A12466.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 769-28-8 | C8H8N2O | 148.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-28-8
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00006269
InChI Key OCYMJCILWYHKAU-UHFFFAOYSA-N
Synonym 2-hydroxy-4,6-dimethylnicotinonitrile, 3-cyano-4,6-dimethyl-2-hydroxypyridine, 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, unii-ots4s2r79e, 4,6-dimethyl-2-hydroxynicotinonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, 3-cyano-2-hydroxy-4,6-dimethylpyridine, nicotinonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, ots4s2r79e
PubChem CID 69856
IUPAC Name 4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile
SMILES CC1=CC(=C(C(=O)N1)C#N)C

Specifications

Melting Point ∼290°C (decomposition)
Quantity 100 g
Beilstein 130992
Formula Weight 148.17
Percent Purity 98%
Chemical Name or Material 3-Cyano-2-hydroxy-4,6-dimethylpyridine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.