missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%

Product Code. 15477196
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477196 1 g 1g
15487196 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477196 Supplier Thermo Scientific Alfa Aesar Supplier No. H62245.03
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 775351-57-0 | C13H15BFNO2 | 247.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775351-57-0
Molecular Formula C13H15BFNO2
Molecular Weight (g/mol) 247.076
MDL Number MFCD06795681
InChI Key RYJOVQGRIOURGT-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-4-fluorophenylboronic acid, pinacol ester, 3-cyano-4-fluorophenylboronic acid pinacol ester, benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyano-4-fluorobenzeneboronic acid pinacol ester, 3-cyano-4-fluorophenyl-boronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
PubChem CID 11402361
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N

Specifications

Melting Point 75°C to 76°C
Quantity 1 g
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Cyano-4-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.