missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10443384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g, Glass bottle
Product Code.
10443384 5g, Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10443384

Brand: Thermo Scientific Acros 398780050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 351003-23-1
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3ClFNO2S
Linear Formula FC6H3(CN)SO2Cl
Quantity 5g
Synonym 4-fluoro-3-cyanobenzenesulfonyl chloride, 3-cyano-4-fluorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-cyano-4-fluoro, 3-cyano-4-fluorobenzenesulphonyl chloride, 3-cyano-4-fluoro-benzenesulfonyl chloride, 4-fluoro-3-cyanobenzenesulfonylchloride, 4-fluoro-3-cyanobenzenesulphonyl chloride, 5-chlorosulfonyl-2-fluorobenzenecarbonitrile, pubchem5110, acmc-20a3ve
InChI Key KDVZADPGGPBAAK-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C#N)F
IUPAC Name 3-cyano-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol) 219.62
PubChem CID 2778908
Formula Weight 219.62
Percent Purity 97%
Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Cyano-4-fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.